Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001025093.2 | NP_001020264.1 | ||||
NM_001284369.1 | NP_001271298.1 | |||||
NM_001284370.1 | NP_001271299.1 | |||||
NM_001284371.1 | NP_001271300.1 | |||||
NM_001284372.1 | NP_001271301.1 | |||||
NM_001284373.1 | NP_001271302.1 | |||||
NM_001284374.1 | NP_001271303.1 | |||||
NM_001284376.1 | NP_001271305.1 | |||||
NM_009715.3 | NP_033845.1 | |||||
GenBank mRNA | AF483482.1 | - | ||||
AF483483.1 | - | |||||
AK035438.1 | - | |||||
AK041145.1 | - | |||||
AK046184.1 | - | |||||
AK047405.1 | - | |||||
AK051591.1 | - | |||||
AK087424.1 | - | |||||
AK139515.1 | - | |||||
AK139844.1 | - | |||||
AK140855.1 | - | |||||
AK144977.1 | - | |||||
AK145887.1 | - | |||||
AK146839.1 | - | |||||
AK218615.1 | - | |||||
AY902311.1 | - | |||||
BC042210.1 | - | 4018323 | ||||
BC079883.1 | - | 6839572 | ||||
BC082596.1 | - | 30649207 | ||||
BC085137.1 | - | |||||
BY474294.1 | - | |||||
M31629.1 | - | |||||
M77167.1 | - | |||||
S76657.1 | - | |||||
S76659.1 | - | |||||
U46026.1 | - |