Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Insr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_010568.2 | NP_034698.2 | ||||
GenBank mRNA | AK052187.1 | - | ||||
AK081856.1 | - | |||||
AK085344.1 | - | |||||
AK164267.1 | - | |||||
AK166467.1 | - | |||||
AK205131.1 | - | |||||
BC029772.1 | - | |||||
BY733217.1 | - | |||||
CR517528.1 | - | |||||
EU346716.1 | - | |||||
J05149.1 | - | |||||
KC356871.1 | - |