Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd226 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001039149.1 | NP_001034238.1 | ||||
NM_178687.2 | NP_848802.2 | |||||
GenBank mRNA | AB195429.1 | - | ||||
AF416980.1 | - | |||||
AF421198.1 | - | |||||
AF421199.1 | - | |||||
AF421200.1 | - | |||||
AK036455.1 | - | |||||
AK140349.1 | - | |||||
BC051526.1 | - | |||||
BC132422.1 | - | 40130699 | ||||
BC138769.1 | - | 8861791 | ||||
BC145410.1 | - | 9053968 | ||||
BC145411.1 | - | |||||
BX526793.1 | - |