Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prrc2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001159634.1 | NP_001153106.1 | ||||
NM_172661.3 | NP_766249.2 | |||||
GenBank mRNA | AK052511.1 | - | ||||
AK077775.1 | - | |||||
AK122300.1 | - | |||||
AK147271.1 | - | |||||
AK147677.1 | - | |||||
AK161103.1 | - | |||||
BC018489.1 | - | |||||
BC038628.1 | - | |||||
BC055116.1 | - | 6329995 | ||||
BP753788.1 | - |