Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il4r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_133380.2 | NP_596871.2 | ||||
XM_006230215.2 | XP_006230277.1 | |||||
XM_006230216.2 | XP_006230278.1 | |||||
XM_008759862.1 | XP_008758084.1 | |||||
XM_008759863.1 | XP_008758085.1 | |||||
XM_008759864.1 | XP_008758086.1 | |||||
GenBank mRNA | AB015746.1 | - | ||||
AB015747.1 | - | |||||
AW527353.1 | - | |||||
BQ204155.1 | - | |||||
X69903.1 | - |