Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: cl9751_1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001146842.1 | NP_001140314.1 | ||||
GenBank mRNA | BT038555.1 | - | ||||
BT068608.1 | - |