Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: polr2k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001016509.1 | NP_001016509.1 | ||||
XM_012964704.2 | XP_012820158.1 | |||||
XM_012964705.2 | XP_012820159.1 | |||||
XM_018094653.1 | XP_017950142.1 | |||||
GenBank mRNA | BC170861.1 | - | ||||
BC170863.1 | - | |||||
CR761743.2 | - | |||||
CR926237.1 | - |