Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100343.3 | NP_171958.1 | 2 | 826 | 107 | |
GenBank mRNA | AB020975.1 | - | 2 | 769 | 107 | |
AF428288.1 | - | 2 | 740 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Both primers and probe map within a single exon |