Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lem-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060138.4 | NP_492539.2 | 4 - 5 | 1822 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |