Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glb-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063981.4 | NP_496382.2 | 4 - 5 | 629 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |