Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbc-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027263.7 | NP_001022434.2 | 7 - 8 | 1454 | 72 | |
NM_001027264.7 | NP_001022435.2 | 7 - 8 | 1456 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |