Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027405.6 | NP_001022576.1 | 7 - 8 | 1251 | 74 | |
GenBank mRNA | U67966.1 | - | 6 - 7 | 1197 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |