Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srh-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072170.2 | NP_504571.2 | 1 - 2 | 593 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |