Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acr-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076727.9 | NP_509128.1 | 2 - 3 | 525 | 84 | |
GenBank mRNA | X86403.1 | - | 404 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |