Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hes1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007648368.1 | XP_007646558.1 | 3 - 4 | 555 | 78 | |
GenBank mRNA | JI894162.1 | - | 3 - 4 | 559 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |