Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMUR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005683202.3 | XP_005683259.1 | 2 - 3 | 906 | 103 | |
GenBank mRNA | FJ842857.1 | - | 2 - 3 | 462 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Capra hircus Probe spans exons |