Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Peritrophin-A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078694.3 | NP_523418.1 | 4 - 5 | 594 | 76 | |
NM_167694.2 | NP_728324.1 | 3 - 4 | 704 | 76 | ||
GenBank mRNA | AF030895.1 | - | 1030 | 76 | ||
AY069467.1 | - | 4 - 5 | 564 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |