Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NitFhit | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080383.4 | NP_525122.1 | 2 | 1419 | 74 | |
GenBank mRNA | AF069989.1 | - | 2 | 1385 | 74 | |
AY089221.1 | - | 2 | 1450 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |