Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs01009663_m1
See other SGIP1 GE Assays ›
Gene Symbol:
SGIP1
Gene Name:
SH3 domain GRB2 like endophilin interacting protein 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 66533272 - 66751139 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs01009663_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: SGIP1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001308203.1
NP_001295132.1
4 - 5
321
117
XM_005271268.3
XP_005271325.1
3 - 4
329
117
XM_005271270.4
XP_005271327.1
4 - 5
660
117
XM_006710969.2
XP_006711032.1
3 - 4
329
117
XM_006710974.2
XP_006711037.1
3 - 4
329
117
XM_011542293.1
XP_011540595.1
3 - 4
329
117
XM_017002514.1
XP_016858003.1
3 - 4
329
117
XM_017002515.1
XP_016858004.1
3 - 4
329
117
XM_017002516.1
XP_016858005.1
4 - 5
452
117
XM_017002518.1
XP_016858007.1
4 - 5
451
117
XM_017002519.1
XP_016858008.1
4 - 5
476
117
XM_017002520.1
XP_016858009.1
3 - 4
329
117
XM_017002521.1
XP_016858010.1
3 - 4
329
117
XM_017002522.1
XP_016858011.1
4 - 5
451
117
XM_017002523.1
XP_016858012.1
4 - 5
475
117
XM_017002528.1
XP_016858017.1
4 - 5
445
117
XM_017002532.1
XP_016858021.1
3 - 4
329
117
XM_017002534.1
XP_016858023.1
3 - 4
329
117
XM_017002535.1
XP_016858024.1
4 - 5
443
117
XM_017002536.1
XP_016858025.1
3 - 4
329
117
XM_017002537.1
XP_016858026.1
4 - 5
442
117
GenBank mRNA
AK299941.1
-
3 - 4
340
117
BX640813.1
-
4 - 5
298
117
Back To Top
More Information
Set Membership:
Amplicon length greater than or equal to 101
Homo sapiens
Probe spans exons
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
endocytosis
positive regulation of receptor-mediated endocytosis
membrane tubulation
protein binding
phospholipid binding
microtubule binding
SH3 domain binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-67fd5c5bdd-q6sk2:80/10.239.167.246:80.
git-commit: 7fb76fe7d0d673187a4c374cd16a9abd242c4866
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.3-offline