Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PCSK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC043562.1 | - | 1 - 2 | 308 | 67 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | small nuclear ribonucleoprotein polypeptide A' | SNRPA1 | Lea1 | -142.0 | Anti-sense | Chr. 15 - 101281510 - 101295282 on NCBI Build GRCh38 |
Downstream | uncharacterized LOC100507472 | LOC100507472 | 1.5kb | Anti-sense | Chr. 15 - 101307251 - 101309304 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons |