Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mybpc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008653.2 | NP_032679.2 | 15 - 16 | 1393 | 57 | |
GenBank mRNA | AF097333.1 | - | 15 - 16 | 1344 | 57 | |
AK146661.1 | - | 15 - 16 | 1394 | 57 | ||
AK146886.1 | - | 15 - 16 | 1394 | 57 | ||
AK169302.1 | - | 14 - 15 | 1371 | 57 | ||
AK169306.1 | - | 14 - 15 | 1371 | 57 | ||
BC094945.1 | - | 15 - 16 | 1390 | 57 | ||
BC131672.1 | - | 15 - 16 | 1369 | 57 | ||
BC154408.1 | - | 15 - 16 | 1414 | 40110890 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |