Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ebf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276387.1 | NP_001263316.1 | 12 - 13 | 2069 | 91 | |
NM_010095.6 | NP_034225.1 | 12 - 13 | 2064 | 91 | ||
GenBank mRNA | AK053013.1 | - | 12 - 13 | 2065 | 91 | |
AK139262.1 | - | 12 - 13 | 1776 | 91 | ||
BC049188.1 | - | 11 - 12 | 1363 | 6312260 | 91 | |
BC050922.1 | - | 11 - 12 | 1586 | 6409737 | 91 | |
U71189.1 | - | 11 - 12 | 1249 | 91 | ||
U82441.1 | - | 11 - 12 | 1118 | 91 | ||
U92703.1 | - | 11 - 12 | 1469 | 91 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |