Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK148184.1 | - | 6 - 7 | 1305 | 78 | |
M84324.1 | - | 6 - 7 | 1052 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |