Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008306.4 | NP_032332.2 | 9 - 10 | 2122 | 89 | |
GenBank mRNA | AF049894.1 | - | 9 - 10 | 1968 | 89 | |
AF074926.1 | - | 9 - 10 | 1968 | 89 | ||
AK147282.1 | - | 10 - 11 | 2515 | 89 | ||
BC066098.1 | - | 9 - 10 | 2122 | 30533342 | 89 | |
BC079561.1 | - | 9 - 10 | 2122 | 30533342 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons |