Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glyat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145935.3 | NP_666047.1 | 1 - 2 | 294 | 110 | |
GenBank mRNA | AK039262.1 | - | 1 - 2 | 295 | 110 | |
AK143803.1 | - | 84 | 110 | |||
AK143901.1 | - | 84 | 110 | |||
BC010799.1 | - | 1 - 2 | 69 | 4234793 | 110 | |
BC015294.1 | - | 69 | 4220471 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |