Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181406.3 | NP_852071.2 | 3 - 4 | 241 | 72 | |
GenBank mRNA | AK075721.1 | - | 3 - 4 | 226 | 72 | |
AK156182.1 | - | 3 - 4 | 219 | 72 | ||
AK166986.1 | - | 3 - 4 | 757 | 72 | ||
BC024878.1 | - | 3 - 4 | 221 | 3989666 | 72 | |
BU961647.1 | - | 243 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |