Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139516.1 | NP_001132988.1 | 5 - 6 | 763 | 74 | |
NM_011249.2 | NP_035379.2 | 5 - 6 | 763 | 74 | ||
GenBank mRNA | AK033402.1 | - | 5 - 6 | 757 | 74 | |
AK147064.1 | - | 5 - 6 | 764 | 74 | ||
AK156055.1 | - | 5 - 6 | 742 | 74 | ||
BC060124.1 | - | 5 - 6 | 730 | 6408844 | 74 | |
U27177.1 | - | 5 - 6 | 747 | 74 | ||
U27178.1 | - | 5 - 6 | 747 | 74 | ||
U33320.1 | - | 5 - 6 | 730 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |