Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Btn1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013483.3 | NP_038511.1 | 6 - 7 | 1042 | 79 | |
GenBank mRNA | AK142596.1 | - | 6 - 7 | 949 | 79 | |
BC011497.1 | - | 6 - 7 | 1016 | 3583827 | 79 | |
BC031459.1 | - | 5 - 6 | 940 | 3582574 | 79 | |
S80642.1 | - | 4 - 5 | 768 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |