Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028977.1 | NP_083253.1 | 1 - 2 | 194 | 81 | |
GenBank mRNA | AK018071.1 | - | 1 - 2 | 160 | 81 | |
BC030317.1 | - | 1 - 2 | 113 | 5372009 | 81 | |
CF914556.1 | - | 1 - 2 | 194 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |