Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Snx19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028874.2 | NP_083150.1 | 8 - 9 | 2821 | 77 | |
GenBank mRNA | AK014388.1 | - | 2 - 3 | 268 | 77 | |
AK032107.1 | - | 4 - 5 | 459 | 77 | ||
AK044307.1 | - | 7 - 8 | 1865 | 77 | ||
AK122231.1 | - | 8 - 9 | 3042 | 77 | ||
BC004635.1 | - | 8 - 9 | 1293 | 77 | ||
BC051164.1 | - | 8 - 9 | 3041 | 77 | ||
BC063262.1 | - | 8 - 9 | 2821 | 6405897 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |