Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Micall1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177461.1 | NP_803412.1 | 3 - 4 | 636 | 69 | |
GenBank mRNA | AK129419.1 | - | 3 - 4 | 374 | 69 | |
AK147587.1 | - | 3 - 4 | 637 | 69 | ||
AK147646.1 | - | 3 - 4 | 647 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |