Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2d26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029562.2 | NP_083838.1 | 3 - 4 | 584 | 79 | |
GenBank mRNA | AK004915.1 | - | 3 - 4 | 585 | 79 | |
BC023241.1 | - | 3 - 4 | 573 | 4222538 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |