Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wrap53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144824.2 | NP_659073.1 | 5 - 6 | 1043 | 99 | |
GenBank mRNA | AK077570.1 | - | 5 - 6 | 947 | 99 | |
BC021790.1 | - | 5 - 6 | 1043 | 4220923 | 99 | |
BC069868.1 | - | 6 - 7 | 1087 | 5101220 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |