Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atf7ip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029253.1 | NP_083529.1 | 4 - 5 | 523 | 125 | |
NM_153123.2 | NP_694763.2 | 4 - 5 | 523 | 125 | ||
GenBank mRNA | AK016180.1 | - | 4 - 5 | 525 | 125 | |
AK049401.1 | - | 4 - 5 | 482 | 125 | ||
AK077229.1 | - | 4 - 5 | 483 | 125 | ||
AK139862.1 | - | 4 - 5 | 480 | 125 | ||
AK145634.1 | - | 4 - 5 | 482 | 125 | ||
AY903215.1 | - | 4 - 5 | 507 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |