Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013842.3 | NP_038870.2 | 4 | 853 | 100 | |
GenBank mRNA | AF027963.3 | - | 4 | 527 | 100 | |
AK007724.1 | - | 4 | 514 | 100 | ||
AK145241.1 | - | 4 | 513 | 100 | ||
AK204892.1 | - | 1 | 49 | 100 | ||
BC008153.1 | - | 4 | 468 | 100 | ||
BC029197.1 | - | 4 | 508 | 2648085 | 100 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |