Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDFI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015102701.2 | XP_014958187.2 | 2 - 3 | 278 | 90 | |
XM_015102702.2 | XP_014958188.2 | 2 - 3 | 282 | 90 | ||
XM_015102703.2 | XP_014958189.2 | 2 - 3 | 213 | 90 | ||
XM_015102704.2 | XP_014958190.2 | 2 - 3 | 139 | 90 | ||
XM_027958635.1 | XP_027814436.1 | 2 - 3 | 273 | 90 |