Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC15A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082700.1 | NP_001076169.1 | 11 - 12 | 1011 | 60 | |
XM_017346648.1 | XP_017202137.1 | 10 - 11 | 978 | 60 | ||
GenBank mRNA | U32507.1 | - | 11 - 12 | 1011 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Oryctolagus cuniculus Probe spans exons |