Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028848198.1 | XP_028704031.1 | 572 | 85 | ||
XM_028848199.1 | XP_028704032.1 | 467 | 85 | |||
XM_028848200.1 | XP_028704033.1 | 581 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |