Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnase2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021664.1 | NP_067696.1 | 4 - 5 | 694 | 74 | |
XM_006233464.2 | XP_006233526.1 | 5 - 6 | 771 | 74 | ||
XM_006233465.2 | XP_006233527.1 | 5 - 6 | 798 | 74 | ||
XM_006233466.2 | XP_006233528.1 | 5 - 6 | 801 | 74 | ||
XM_006233468.2 | XP_006233530.1 | 5 - 6 | 1182 | 74 | ||
XM_008761521.1 | XP_008759743.1 | 5 - 6 | 904 | 74 | ||
GenBank mRNA | AF178974.1 | - | 4 - 5 | 694 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Rattus norvegicus Probe spans exons |