Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mefv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031634.1 | NP_113822.1 | 6 - 7 | 1843 | 66 | |
GenBank mRNA | AF143410.1 | - | 6 - 7 | 1843 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |