Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107097.1 | NP_001100567.1 | 5 - 6 | 1960 | 89 | |
XM_006248251.2 | XP_006248313.1 | 5 - 6 | 1636 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus Probe spans exons |