Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UQCRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185172.1 | NP_001172101.1 | 3 - 4 | 289 | 74 | |
XM_021088626.1 | XP_020944285.1 | 249 | 74 | |||
XM_021088627.1 | XP_020944286.1 | 336 | 74 | |||
GenBank mRNA | AK347211.1 | - | 3 - 4 | 289 | 74 | |
AY610483.1 | - | 2 - 3 | 265 | 74 | ||
EV895402.2 | - | 2 - 3 | 216 | 74 | ||
EV897222.2 | - | 3 - 4 | 298 | 74 | ||
EW033712.2 | - | 2 - 3 | 244 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons |