Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BZW1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244493.1 | NP_001231422.1 | 8 - 9 | 902 | 115 | |
XM_021074514.1 | XP_020930173.1 | 1228 | 115 | |||
GenBank mRNA | AK239661.1 | - | 8 - 9 | 908 | 115 | |
AK343867.1 | - | 8 - 9 | 953 | 116 | ||
AK343897.1 | - | 6 - 7 | 748 | 116 | ||
AK345146.1 | - | 2 - 3 | 500 | 115 | ||
AY609907.1 | - | 8 - 9 | 906 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |