Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pPLAIIIbeta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115352.4 | NP_191055.1 | 1 - 2 | 1939 | 103 | |
GenBank mRNA | AK229196.1 | - | 1 - 2 | 1612 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |