Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT3G56130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035794.1 | NP_001030871.1 | 7 - 8 | 1181 | 74 | |
NM_115471.3 | NP_567035.1 | 8 - 9 | 1052 | 74 | ||
NM_202717.1 | NP_974446.1 | 7 - 8 | 1177 | 74 | ||
GenBank mRNA | AF378871.1 | - | 8 - 9 | 928 | 74 | |
AK316707.1 | - | 7 - 8 | 1032 | 74 | ||
AY050468.1 | - | 8 - 9 | 776 | 74 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |