Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYB16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343396.1 | NP_001330733.1 | 2 - 3 | 916 | 98 | |
NM_121535.4 | NP_197035.1 | 3 - 4 | 834 | 98 | ||
GenBank mRNA | AF370613.1 | - | 3 - 4 | 611 | 98 | |
AK228474.1 | - | 3 - 4 | 761 | 98 | ||
AY519624.1 | - | 3 - 4 | 611 | 98 | ||
BT028929.1 | - | 3 - 4 | 611 | 98 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |