Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161190.2 | NP_001154662.2 | 23 - 24 | 4541 | 72 | |
NM_001161191.3 | NP_001154663.2 | 23 - 24 | 4659 | 72 | ||
NM_001339114.1 | NP_001325812.1 | 23 - 24 | 4669 | 72 | ||
NM_001339116.1 | NP_001325813.1 | 18 - 19 | 4042 | 72 | ||
NM_114260.1 | NP_189978.1 | 21 - 22 | 4394 | 72 | ||
GenBank mRNA | DQ479972.1 | - | 21 - 22 | 4394 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |