Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPKKK15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345106.1 | NP_001330365.1 | 1 | 1344 | 72 | |
NM_124891.2 | NP_200320.1 | 1 | 1312 | 72 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |