Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130151.7 | NP_182112.2 | 6 - 7 | 2199 | 131 |
Set Membership: |
![]() ![]() ![]() ![]() |