Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202664.1 | NP_001189593.1 | 3 - 4 | 636 | 99 | |
NM_128025.4 | NP_565576.1 | 2 - 3 | 567 | 99 | ||
GenBank mRNA | AF360315.1 | - | 2 - 3 | 544 | 99 | |
AK319102.1 | - | 3 - 4 | 609 | 99 | ||
AY056366.1 | - | 2 - 3 | 454 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |